Learn More
Gibco™ Human SCF Recombinant Protein
Recombinant Human Stem Cell Factor (SCF, C-Kit Ligand) is a bioactive protein intended for use in cell culture applications
Brand: Gibco™ PHC2113
Code : 50
Additional Details : Weight : 0.50000kg
Description
Human SCF Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
SCF is a pleiotropic factor that acts in utero in germ cell and neural cell development, and hematopoiesis, all believed to reflect a role in cell migration. In adults, it functions pleiotropically, while mostly noted for its continued requirement in hematopoiesis.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of human MO-7e cells
- Proven Compatibility: Gibco proteins are bioassayed with Gibco media
Shipping Condition: Wet Ice.
Specifications
Cell Culture | |
18.54 kDa | |
1mg | |
Human | |
Carrier-Free | |
1 mg | |
E. coli | |
Recombinant, Ligand | |
Other Proteins | |
Gibco™ |
4254 | |
Sequential Chromatography | |
SCF (c-kit Ligand) | |
Store in refrigerator (2–8°C). | |
< 0.1 ng/μg | |
Wet Ice | |
Cytokines & Receptors | |
Lyophilized | |
Stem Cell Research, Oncology | |
95 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.