Learn More
Invitrogen™ CD93 (Soluble) Human Uncoated ELISA Kit with Plates
Description
Includes
- Capture Antibody Concentrate
- Detection Antibody Concentrate
- Human sCD93 Standard (lyophilized): 800ng/mL upon reconstitution
- Phosphate Buffered Saline Concentrate (PBS)
- Assay Buffer A Concentrate (PBS with 1% Tween 20 and 10% BSA)
- Sample Diluent Z
- Detection enzyme
- Tetramethylbenzidine (TMB) Substrate Solution
CD93 (Soluble) HumanUncoated ELISA Kit contains pre-matched antibody pairs, plates, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of rat total IgG. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Specifications
Specifications
| Assay Sensitivity | 3.3ng/mL |
| Sample Type | Cell Culture Supernatant, Plasma, Serum |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Quantity | 10 x 96 Tests |
| Synonym | soluble C1q receptor |
| Target | CD93 |
For Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.