missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD93 (Soluble) Human Uncoated ELISA Kit with Plates

Product Code. 15540867
Click to view available options
Quantity:
10 x 96 Tests
2 x 96 Tests
20 x 96 Tests
Unit Size:
Pack of 10
Pack of 2
Pack of 20
This item is not returnable. View return policy

Product Code. 15540867

Brand: Invitrogen™ 885034076

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

  • Capture Antibody Concentrate
  • Detection Antibody Concentrate
  • Human sCD93 Standard (lyophilized): 800ng/mL upon reconstitution
  • Phosphate Buffered Saline Concentrate (PBS)
  • Assay Buffer A Concentrate (PBS with 1% Tween 20 and 10% BSA)
  • Sample Diluent Z
  • Detection enzyme
  • Tetramethylbenzidine (TMB) Substrate Solution

For quantitative measurement of human sCD93

CD93 (Soluble) HumanUncoated ELISA Kit contains pre-matched antibody pairs, plates, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of rat total IgG. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Human sCD93 ELISA kit contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA).
TRUSTED_SUSTAINABILITY

Specifications

Assay Sensitivity 3.3ng/mL
Sample Type Cell Culture Supernatant, Plasma, Serum
Target Species Human
For Use With (Application) ELISA
Quantity 20 x 96 Tests
Synonym soluble C1q receptor
Target CD93

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.