missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Human sCD44var (v5) Platinum ELISA Kit
For the quantitative detection of human sCD44var (v5).
Brand: Invitrogen™ BMS220TEN
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD44var (v5)
HRP-Conjugate anti-human sCD44var (v5) monoclonal antibody
Human sCD44var (v5) Standard lyophilized, 20ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films
Description
The sCD44var(v5) ELISA detects all circulating CD44 isoforms comprising the sCD44var(v5) sequencesPlatinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications
HRP | |
10 x 96 tests | |
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin), amniotic fluid, urine | |
Human | |
VFF-8 Phagocytic glycoprotein I, Hermes antigen |
ELISA | |
0.14ng/mL | |
20μL (1:6 prediluted) | |
sCD44var (v5) |
For Research Use Only