missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SCAND2 Partial ORF (NP_071333, 1 a.a. - 62 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00054581-Q02.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16005545

  • £425.00 / 25µg
Estimated Shipment: 06-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The SCAN domain is a highly conserved, leucine-rich motif of approximately 60 aa originally found within a subfamily of zinc finger proteins. This gene belongs to a family of genes that encode an isolated SCAN domain, but no zinc finger motif. Functional studies have established that the SCAN box is a protein interaction domain that mediates both hetero- and homoprotein associations, and maybe involved in regulation of transcriptional activity. Multiple transcript variants which encode the same isoform but differ only in their 3' UTRs, and another variant which encodes a distinct isoform have been described for this gene.

Sequence: MAVAVDQQIQTPSVQDLQIVKLEEDSHWEQEISLQGNYPGPETSCQSFWHFRYQEASRPREA
Specifications

Specifications

NP_071333
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.56kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MAVAVDQQIQTPSVQDLQIVKLEEDSHWEQEISLQGNYPGPETSCQSFWHFRYQEASRPREA
RUO
SCAND2
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
54581
SCAND2 (Human) Recombinant Protein (Q02)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SCAND2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers