missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SAT1 (aa 19-56) Control Fragment Recombinant Protein

Product Code. 30201956
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30201956

Brand: Invitrogen™ RP103212

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63107 (PA5-63107. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Spermidine/spermine-N1-acetyltransferase (SSAT1) is a short-lived polyamine catabolic enzyme which catalyzes the acetylation of polyamines. This highly regulated enzyme allows a fine attenuation of the intracellular concentration of polyamines. Induction of SSAT1 plays an important role in polyamine homoeostasis, since the N1-acetylated polyamines can be excreted or oxidized by acetylpolyamine oxidase.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P21673
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6303
Name Human SAT1 (aa 19-56) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AA617398; Ab2-402; CAON; DC21; diamine acetyltransferase 1; diamine acetyltransferase 1; spermidine/spermine N1-acetyltransferase 1; diamine N-acetyltransferase 1; diamine N-acetyltransferase 1 A; EC 2.3.1.57; EDM4; epid34; hypothetical protein LOC508861; kfsd; KFSDX; Polyamine N-acetyltransferase 1; putrescine acetyltransferase; SAT; Sat1; SAT-1; sat1.S; sat1a; sat1-a; sat1-b; Slc26a1; solute carrier family 26 (anion exchanger), member 1; solute carrier family 26 (sulfate transporter), member 1; solute carrier family 26 member 1; solute carrier family 26, member 1; Spermidine / spermine N1-acyltransferase (diamine acetyltransferase); spermidine/spermine N(1)-acetyltransferase 1; spermidine/spermine N1-acetyl transferase 1; spermidine/spermine N1-acetyltransferase; spermidine/spermine N1-acetyltransferase 1; spermidine/spermine N1-acetyltransferase 1 A; spermidine/spermine N1-acetyltransferase 1 S homeolog; spermidine/spermine N1-acetyltransferase alpha; spermidine/spermine N-acetyltransferase; SSAT; SSAT-1; sulfate anion tranporter AT1; sulfate anion transporter 1; sulfate anion transporter-1; sulfate/anion transporter SAT-1 protein; XELAEV_18014544mg
Common Name SAT1
Gene Symbol SAT1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RLIKELAKYEYMEEQVILTEKDLLEDGFGEHPFYHCLV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.