missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SARS2 (aa 104-177) Control Fragment Recombinant Protein

Product Code. 30199606
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30199606

Brand: Invitrogen™ RP101648

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (80%), Rat (80%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84321 (PA5-84321. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes the mitochondrial seryl-tRNA synthetase precursor, a member of the class II tRNA synthetase family. The mature enzyme catalyzes the ligation of Serine to tRNA(Ser) and participates in the biosynthesis of selenocysteinyl-tRNA(sec) in mitochondria. The enzyme contains an N-terminal tRNA binding domain and a core catalytic domain. It functions in a homodimeric form, which is stabilized by tRNA binding. This gene is regulated by a bidirectional promoter that also controls the expression of mitochondrial ribosomal protein S12. Both genes are within the critical interval for the autosomal dominant deafness locus DFNA4 and might be linked to this disease. Multiple transcript variants encoding different isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NP81
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 54938
Name Human SARS2 (aa 104-177) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2410015F05Rik; D7Ertd353e; EC 6.1.1.11; mitochondrial seryl-tRNA synthetase; mtSerRS; SARS; Sars2; Sarsm; serine tRNA ligase 2, mitochondrial; serine-tRNA ligase; serine-tRNA ligase, mitochondrial; serine--tRNA ligase, mitochondrial; SerRS; SerRS2; SerRSmt; SERS; seryl-aminoacyl-tRNA synthetase 2; Seryl-tRNA synthetase; seryl-tRNA synthetase 2; seryl-tRNA synthetase 2, mitochondrial; seryl-tRNA synthetase, mitochondrial; seryl-tRNA(Ser/Sec) synthetase; SYS; zgc:66345
Common Name SARS2
Gene Symbol SARS2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RSLEEEKAAVTEAVRALLANQDSGEVQQDPKYQGLRARGREIRKELVHLYPREAQLEEQFYLQALKLPNQTHPD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.