missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SARS-CoV-2 Spike (Trimer) IgM ELISA Kit

Product Code. 17106398
Click to view available options
Quantity:
10 x 96 Tests
2 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 2
Pack of 96
This item is not returnable. View return policy

Product Code. 17106398

Brand: Invitrogen™ BMS2324

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

It has recently been shown that SARS (severe acute respiratory syndrome) is caused by a human coronavirus. Human coronaviruses are the major cause of upper respiratory tract illness, such as the common cold, in humans. Coronaviruses are positive-stranded RNA viruses, featuring the largest viral RNA genomes known to date (27-31 kb). The first step in coronavirus infection is binding of the viral spike protein, a 139-kDa protein, to certain receptors on host cells. The spike protein is the main surface antigen of the coronavirus. The glycosylated spike protein (as well as the nucleocapsid protein) can be detected in infected cell culture supernatants with antisera from SARS patients.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 46.88 to 3,000 Units/mL
Assay Sensitivity <234 Units/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum
For Use With (Equipment) Colorimetric Microplate Reader
Gene Alias Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV-2
Interassay CV <10%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, High Control (standard), Calibrator (medium control), Low Control, IgM - HRP conjugate, Assay Buffer concentrate, Wash Buffer concentrate, Substrate solution, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 5 μL, Serum 5 μL
Storage Requirements 2°C to 8°C
Target SARS-CoV-2 Spike (trimer) Protein
Test Time 30 mins
Total Assay Time 1 hrs 15 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.