missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SALM2/LRFN1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5669-SP
This item is not returnable.
View return policy
Description
SALM2/LRFN1 Monoclonal specifically detects SALM2/LRFN1 in Human samples. It is validated for Immunohistochemistry.
Specifications
| SALM2/LRFN1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| KIAA1484, leucine rich repeat and fibronectin type III domain containing 1, leucine-rich repeat and fibronectin type III domain-containing protein 1, LRFN1, SALM2, synaptic adhesion-like molecule 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 57622 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Immunohistochemistry | |
| 610712 | |
| Immunohistochemistry 8-25 ug/mL | |
| Q9P244 | |
| LRFN1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human SALM2 Gln32-Gly534 (predicted) Accession # Q9P244 | |
| 25 μg | |
| Primary | |
| Detects human SALM2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human SALM1, 3, or 4 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction