missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SACS Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF8014-SP
This item is not returnable.
View return policy
Description
SACS Polyclonal specifically detects SACS in Human samples. It is validated for Immunohistochemistry.
Specifications
| SACS | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ARSACS, dnaJ homolog subfamily C member 29, DNAJC29, KIAA0730, Sacsin, spastic ataxia of Charlevoix-Saguenay (sacsin) | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 26278 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Immunohistochemistry | |
| LYOPH | |
| Immunohistochemistry 5-15 ug/mL | |
| Q9NZJ4 | |
| SACS | |
| E. coli-derived recombinant human SACS Asn4402-Val4579 Accession # Q9NZJ4 | |
| 25 μg | |
| Primary | |
| Detects human SACS in direct ELISAs. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction