missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human S1P3/EDG-3 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB7524U-100UG
This item is not returnable.
View return policy
Description
S1P3/EDG-3 Monoclonal specifically detects S1P3/EDG-3 in Human samples. It is validated for Flow Cytometry.
Specifications
| S1P3/EDG-3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q99500 | |
| S1PR3 | |
| Human S1P3/EDG-3 transfected NS0 cells Accession # Q99500 | |
| RUO | |
| 1903 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 776808 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| EDG3, EDG-3, EDG3FLJ37523, Endothelial differentiation G-protein coupled receptor 3, endothelial differentiation, sphingolipid G-protein-coupled receptor, 3, FLJ93220, G protein-coupled receptor, endothelial differentiation gene-3, LPB3, MGC71696, S1P receptor 3, S1P receptor Edg-3, S1P receptor EDG3, S1P3, S1PR3, sphingosine 1-phosphate receptor 3, Sphingosine 1-phosphate receptor Edg-3, sphingosine-1-phosphate receptor 3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human S1P 3EDG-3 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction