missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human S100G (aa 5-59) Control Fragment Recombinant Protein

Product Code. 30210419
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210419

Brand: Invitrogen™ RP101788

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (80%), Rat (80%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63195 (PA5-63195. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

S100G encodes calbindin D9K, a vitamin D-dependent calcium-binding protein. This cytosolic protein belongs to a family of calcium-binding proteins that includes calmodulin, parvalbumin, troponin C, and S100 protein. In the intestine, the protein is vitamin D-dependent and its expression correlates with calcium transport activity. The protein may increase Ca2+ absorption by buffering Ca2+ in the cytoplasm and increase ATP-dependent Ca2+ transport in duodenal basolateral membrane vesicles.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P29377
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 795
Name Human S100G (aa 5-59) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 9 kDa CaBP; AU042539; CABP; CABP1; CABP9K; CaBP-D9K; Calb3; calbindin 3, (vitamin D-dependent calcium binding protein); calbindin 3, (vitamin D-dependent calcium-binding protein); calbindin D9k; calbindin-D9K; calbindin-D9K major form; calcium-binding protein, intestinal, vitamin D-dependent (9-kDa CaBP); Cbpi; cholecalcin; cytidine 5'-triphosphate synthase 2; protein S100-G; Rncalbd9; S100 calcium binding protein G; S100 calcium-binding protein G; S100D; S100g; Vitamin D-dependent calcium-binding protein, intestinal
Common Name S100G
Gene Symbol S100G
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KSPEELKRIFEKYAAKEGDPDQLSKDELKLLIQAEFPSLLKGPNTLDDLFQELDK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.