missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human S100B Recombinant Protein
Description
This product contains purified recombinant human S100 beta Chain protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.
S100B is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells and are involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100B may function in neurite extension, proliferation of melanoma cells, stimulation of Ca2+ fluxes, inhibition of PKC-mediated phosphorylation, astrocytosis and axonal proliferation, and inhibition of microtubule assembly. Chromosomal rearrangements and altered expression of this gene have been implicated in several diseases, including Alzheimer's disease, Down's syndrome, epilepsy, amyotrophic lateral sclerosis, melanoma, and type I diabetes.
Specifications
Specifications
| Accession Number | P04271 |
| Concentration | 0.5 mg/mL |
| For Use With (Application) | Control, Western Blot |
| Formulation | PBS with 50% glycerol and no preservative; pH 7.4 |
| Gene ID (Entrez) | 6285 |
| Name | Human S100B |
| pH Range | 7.4 |
| Quantity | 100 μg |
| Source | E. coli |
| Immunogen | N/A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?