missing translation for 'onlineSavingsMsg'
Learn More

Human S100A8/S100A9 Heterodimer Alexa Fluor 750-conjugated Antibody, R&D Systems™

Product Code. 18701167
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18701167 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18701167 Supplier R&D Systems Supplier No. IC45703S100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

S100A8/S100A9 Heterodimer Monoclonal specifically detects S100A8/S100A9 Heterodimer in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100A8/S100A9 Heterodimer
Applications Flow Cytometry
Classification Monoclonal
Clone 900028
Conjugate Alexa Fluor 750
Dilution Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P05109 (S100A8) and P06702 (S100A9)
Host Species Mouse
Immunogen E. coli-derived recombinant human S100A8/S100A9 heterodimer, Met1-Glu93 (S100A8) & Thr2-Pro114 (S100A9), Accession No. P05109 (S100A8) and P06702 (S100A9)
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Test Specificity Detects human S100A8/S100A9 heterodimer in direct ELISAs. In direct ELISAs, no detection of recombinant human S100A8 or S100A9 monomers was observered.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.