missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human S100A8/S100A9 Heterodimer Alexa Fluor 647-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems IC9337R-025
This item is not returnable.
View return policy
Description
S100A8/S100A9 Heterodimer Monoclonal specifically detects S100A8/S100A9 Heterodimer in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| S100A8/S100A9 Heterodimer | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Detects human S100A8/S100A9 in direct ELISAs. In ELISAs, it detects recombinant human S100A8/S100A9 heterodimer but it does not detect recombinant human S100A8 or S100A9 monomers. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Flow Cytometry | |
| 900028 | |
| Intracellular Staining by Flow Cytometry 5 uL/10^6 cells | |
| P05109 (S100A8) & P06702 (S100A9) | |
| E. coli-derived recombinant human S100A8/S100A9 heterodimer Met1-Glu93 (S100A8) & Thr2-Pro114 (S100A9) Accession # P05109 (S100A8) & P06702 (S100A9) | |
| 25 Tests | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction