missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human S100A7 Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 1 publication
Brand: R&D Systems AF4475
Additional Details : Weight : 0.00970kg
Description
S100A7/Psoriasin Polyclonal specifically detects S100A7/Psoriasin in Human samples. It is validated for Western Blot.Specifications
S100A7/Psoriasin | |
Polyclonal | |
Unconjugated | |
AAA60210 | |
S100A7 | |
E. coli-derived recombinant human S100A7 Met1-Gln101 Accession # AAA60210 | |
100 μg | |
Primary | |
Detects human S100A7 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human S100A8 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
HID 5, HID5, HID-5, protein S100-A7, PSOR1, Psoriasin, psoriasin 1, S100 calcium binding protein A7, S100 calcium binding protein A7 (psoriasin 1), S100 calcium-binding protein A7, S100 calcium-binding protein A7 (psoriasin 1), S100A7c | |
Sheep | |
Affinity Purified | |
RUO | |
6278 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only