missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human S100A11 Recombinant Protein

Product Code. 11589031
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11589031 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11589031 Supplier Invitrogen™ Supplier No. RP75719

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This product contains purified recombinant human S100A11 protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in motility, invasion, and tubulin polymerization. Chromosomal rearrangements and altered expression of this gene have been implicated in tumor metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P31949
For Use With (Application) Control, Western Blot
Formulation 20 mM tris HCl with 50% glycerol, 5 mM EDTA and no preservative; pH 8
Gene ID (Entrez) 6282
Name Human S100A11
Quantity 100 μg
Source E. coli
Immunogen N/A
Regulatory Status RUO
Gene Alias cal; Calgizzarin; calizzarin; EMAP; Emap1; EMAPI; endothelial monocyte activating polypeptide 1; endothelial monocyte-activating polypeptide; epididymis secretory protein Li 43; HEL-S-43; Metastatic lymph node gene 70 protein; MLN 70; MLN70; Protein S100-A11; Protein S100-A11, N-terminally processed; Protein S100C; Protein S100-C; S100 calcium binding protein A11; S100 calcium binding protein A11 (calgizzarin); S100 calcium binding protein A11 (calizzarin); S100 calcium binding protein A14; S100 calcium-binding protein A11; S100 calcium-binding protein A11 (calgizzarin); S100a11; S100a14; S100C
Common Name S100A11
Gene Symbol S100A11
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Met1-Thr105 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.