missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human S100A11 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF4874-SP
This item is not returnable.
View return policy
Description
S100A11 Polyclonal specifically detects S100A11 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| S100A11 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| calgizzarin, Metastatic lymph node gene 70 protein, MLN 70, MLN70, protein S100-A11, Protein S100-C, S100 calcium binding protein A11, S100 calcium-binding protein A11, S100 calcium-binding protein A11 (calgizzarin), S100C, S100CS100 calcium binding protein A11 (calgizzarin) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6282 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| P31949 | |
| S100A11 | |
| E. coli-derived recombinant human S100A11 Ala2-Thr105 Accession # P31949 | |
| 25 μg | |
| Primary | |
| Detects human S100A11 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) S100A1, A2, A7, A8, A10, A13, A16, rhS100B, rhS100P, recombinant mouse (rm) S100A6, and rmS100A10 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction