missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human S100A10 Recombinant Protein

Product Code. 11549031
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11549031 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11549031 Supplier Invitrogen™ Supplier No. RP75715

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This product contains purified recombinant human S100A10 protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.

The S-100 family of calcium-activated proteins interact with a range of target proteins to modulate biological signaling pathways. Numerous cancer cell lines overexpress the plasminogen receptor S-100A10 on the extracellular cell surface, where it forms a heterotetrameric complex with Annexin II, though this association is not required for plasma membrane localization or binding and activation of plasminogen. Additionally, S-100A10 acts as a cellular chaperone for hepatitis B (Hep B) virus polymerase. Hep B virus polymerase normally localizes to the cytoplasm only, though in the presence of S-100A10 a portion relocates to the nucleus, implying a role for S-100A10 and intracellular calcium in the process of viral replication.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P60903
Concentration 0.10 mg/mL
For Use With (Application) Control, Western Blot
Formulation PBS with 50% glycerol and no preservative
Gene ID (Entrez) 6281
Molecular Weight (g/mol) 15.3 kDa
Name Human S100A10
Quantity 10 μg
Source E. coli
Immunogen N/A
Regulatory Status RUO
Gene Alias 42 C; AA409961; AL024248; Annexin II; annexin II ligand; annexin II ligand, calpactin I, light polypeptide; annexin II tetramer (AIIt) p11 subunit; AN x 2 L; AN x 2 LG; Ca[1 ]; CAL12; Cal1l; calcium binding protein A11 (calgizzarin); calpactin I light chain; Calpactin-1 light chain; cellular ligand of annexin II; CLP11; GP11; MGC111133; MGC133268; Nerve growth factor-induced protein 42 C; OTTHUMP00000015270; P PRSS26; p10; p10 protein; P11; p11 subunit; Protein S100 A10; Protein S100-A10; S100 calcium binding protein A10; S100 calcium binding protein A10 (annexin II ligand, calpactin I, light polypeptide (p11)); S100 calcium binding protein A10 (calgizzarin); S100 calcium binding protein A10 (calpactin); S100 calcium-binding protein A10; S100 calcium-binding protein A10 (annexin II ligand, calpactin I, light polypeptide (p11)); S-100 related protein, clone 42 C; S100a10
Common Name S100A10
Gene Symbol S100A10
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Met1-Lys97 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.