missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RWDD2B (aa 252-319) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53778 (PA5-53778. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Accession Number | P57060 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 10069 |
| Name | Human RWDD2B (aa 252-319) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | C21orf6; GL011; RWD domain containing 2 B; RWD domain-containing protein 2 B; RWDD2B |
| Common Name | RWDD2B |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction