missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RSPH6A (aa 114-190) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (46%), Rat (46%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60986 (PA5-60986. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The protein encoded by this gene is similar to a sea urchin radial spoke head protein. Radial spoke protein complexes form part of the axoneme of eukaryotic flagella and are located between the axoneme's outer ring of doublet microtubules and central pair of microtubules. In Chlamydomonas, radial spoke proteins are thought to regulate the activity of dynein and the symmetry of flagellar bending patterns. This gene maps to a region of chromosome 19 that is linked to primary ciliary dyskinesia-2 (CILD2).
Specifications
Specifications
| Accession Number | Q9H0K4 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 81492 |
| Name | Human RSPH6A (aa 114-190) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | Dmwd; dystrophia myotonica-containing WD repeat motif; hematological and neurological expressed 1 (Hn1-ps1) pseudogene; Hn1-ps1; ortholog of mouse radial spokehead-like 1; radial spoke head 6 homolog A; radial spoke head 6 homolog A (Chlamydomonas); radial spoke head protein 6 homolog A; Radial spoke head-like protein 1; radial spokehead-like 1; Rshl1; RSP4; RSP6; RSPH4B; RSPH6A |
| Common Name | RSPH6A |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction