missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RSG1 (aa 83-175) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65309 (PA5-65309. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Potential effector of the planar cell polarity signaling pathway. Plays a role in targeted membrane trafficking most probably at the level of vesicle fusion with membranes. Involved in cilium biogenesis by regulating the transport of cargo proteins to the basal body and to the apical tips of cilia. More generally involved in exocytosis in secretory cells.
Specifications
Specifications
| Accession Number | Q9BU20 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 79363 |
| Name | Human RSG1 (aa 83-175) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 6330545A04Rik; C1orf89; Ciliogenesis and planar polarity effector 2; ciliogenesis and planar polarity effector 2; REM2- and Rab-like small GTPase 1; CPLANE2; Gm723; im:7136213; miro domain-containing protein C1orf89; miro domain-containing protein C1orf89 homolog; Rem/Rab-Similar GTPase 1; REM2 and RAB like small GTPase 1; REM2- and Rab-like small GTPase 1; REM2 and RAB-like small GTPase 1; RGD1561421; RSG1; zgc:101035; zgc:101035 protein; zgc:113864 |
| Common Name | RSG1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction