missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RRBP1 (aa 1055-1178) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (77%), Rat (77%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82444 (PA5-82444. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Analysis of cDNA clones indicates that ribosome binding protein 1 may exist in different forms due to removal of tandem repeats, or partial intraexonic splicing of RRBP1. The form presented here is lacking the canine p180 ribosome-binding domain, NQGKKAEGAQ, which is tandemly repeated close to the N-terminus in other forms that haven't been fully characterized. RRBP1 has been excluded as a candidate gene in the cause of Alagille syndrome. Alternate splicing results in multiple transcript variants.
Specifications
Specifications
| Accession Number | Q9P2E9 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 6238 |
| Name | Human RRBP1 (aa 1055-1178) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1700087N07Rik; 180 kDa ribosome receptor homolog; 5730465C04Rik; ES/130; ES/130-related protein; ES130; hES; KIAA1398; mKIAA1398; mRRp; mRRp0; mRRp1.8; mRRp10; mRRp15a; mRRp15b; mRRp16.8; mRRp2; mRRp41; mRRp47; mRRp5.4; p180; ribosome binding protein 1; ribosome binding protein 1 homolog 180 kDa (dog); ribosome receptor protein; Ribosome-binding protein 1; RP11-462D18.3; Rrbp1; RRp |
| Common Name | RRBP1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction