missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RPUSD2 (aa 278-359) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59856 (PA5-59856. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The function remains unknown.
Specifications
Specifications
| Accession Number | Q8IZ73 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 27079 |
| Name | Human RPUSD2 (aa 278-359) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 4921503C21Rik; 9630001E10; BB231107; C15orf19; C18B11; C18B11 homolog (44.9 kD); CG6187-like; FLJ31409; RGD1306147; RNA pseudouridine synthase domain containing 2; RNA pseudouridylate synthase domain containing 2; RNA pseudouridylate synthase domain-containing protein 2; Rpusd2 |
| Common Name | RPUSD2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction