missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RPS21 (aa 1-81) Control Fragment Recombinant Protein

Product Code. 30210955
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210955

Brand: Invitrogen™ RP102214

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51914. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Differentiated embryo-chondrocyte expressed gene 1 (DEC1) is an important protein involved in embryonic cell differentiation, proliferation, and apoptosis. It has also recently been shown to be regulated by hypoxia. The loss of DEC1 expression may be an early event in the development of lung cancer, while DEC1 may be up-regulated by hypoxia in other cancers and in more extreme hypoxia it may have a role in cell death.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P63220
Concentration 2.9 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 6227
Name Human RPS21 (aa 1-81) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 1810049N11Rik; 2410030A14Rik; 40S ribosomal protein S21; 8.2 kDa differentiation factor; HLDF; human leukemia differentiation factor; LOC615178 protein; ribosomal protein S21; rps21; S21; Small ribosomal subunit protein eS21; zgc:56642; zgc:86816
Common Name RPS21
Gene Symbol RPS21
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MQNDAGEFVDLYVPRKCSASNRIIGAKDHASIQMNVAEVDKVTGRFNGQFKTYAICGAIRRMGESDDSILRLAKADGIVSK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.