missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RPS17 (aa 67-108) Control Fragment Recombinant Protein

Product Code. 30213013
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30213013

Brand: Invitrogen™ RP100843

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63059 (PA5-63059. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron-containing gene within their coding regions. Ribosomal Protein S17, also known as RPS17, RPS17L1 or RPS17L2, is a 135 amino acid protein that is a component of the 40S subunit. Localized to the cytoplasm and expressed ubiquitously, Ribosomal Protein S17 belongs to the S17e family of ribosomal proteins and functions in protein synthesis. Mutations in the gene encoding Ribosomal Protein S17 are associated with Diamond-Blackfan anemia (DBA), a rare congenital disorder characterized by defective differentiation of pro-erythroblasts. Like most ribosomal proteins, Ribosomal Protein S17 exists as multiple processed pseudogenes that are scattered throughout the genome.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08708
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6218
Name Human RPS17 (aa 67-108) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 40 S ribosomal protein S17; DBA4; ribosomal protein S17; Rps17; RPS17L; RPS17L1; RPS17L2; S17; Small ribosomal subunit protein eS17
Common Name RPS17
Gene Symbol Rps17
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RGISIKLQEEERERRDNYVPEVSALDQEIIEVDPDTKEMLKL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.