missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RPP38 (aa 189-277) Control Fragment Recombinant Protein

Code produit. 30207381
Click to view available options
Quantity:
100 μL
Conditionnement:
100µL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 30207381

Marque: Invitrogen™ RP100663

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (75%), Rat (75%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111212 (PA5-111212. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5'-ends. RPP38 may associate transiently with RNase P RNA as a factor involved in the transport of H1 RNA to the putative site of its assembly in the cell, the nucleolus.
TRUSTED_SUSTAINABILITY

Spécification

Accession Number P78345
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10557
Name Human RPP38 (aa 189-277) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C330006A15Rik; ribonuclease P protein subunit p38; ribonuclease P/MRP 38 subunit; ribonuclease P/MRP 38 kDa subunit; ribonuclease P/MRP subunit p38; RNaseP protein p38; RPP38
Common Name RPP38
Gene Symbol RPP38
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FVDEVRAIIPRVPSLSVPWLQDRIEDSGENLETEPLESQDRELLDTSFEDLSKPKRKLADGRQASVTLQPLKIKKLIPNPNKIRKPPKS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.