missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Human RPLPO (Large Ribosomal Protein) Endogenous Control, VIC™/MGB probe, primer limited
GREENER_CHOICE

Product Code. 10638685 Shop All Applied Biosystems Products
Change view
Click to view available options
2500 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10638685 2500 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10638685 Supplier Applied Biosystems™ Supplier No. 4326314E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Human RPLPO (Large Ribosomal Protein) Endogenous Control, VIC™/MGB probe, primer limited

Human RPLPO (Large Ribosomal Protein) Endogenous Control allows relative gene expression quantification in cDNA samples when used with other gene expression assays. The probe is labeled with VIC™ dye-MGB, and the primers are limited. It can be used for multiplex or singleplex PCR reactions. This endogenous control is designed to be used with TaqMan™ gene expression assays or TaqMan™ primers/probes.

Assay Details:

Gene Symbol: RPLPO
RefSeq: NM_001002.3
Probe Exon Location:3
Amplicon Size: 105
Corresponding TaqMan Assay ID: Hs99999902_m1

TaqMan™ Endogenous Controls

Eliminate months of assay design, formulation, and testing by using TaqMan™ endogenous controls to quantify gene expression. This convenient collection of pre-designed primer/probe sets enables you to normalize the amount of sample RNA or DNA in a reaction.

Complete Solution for Quantitative Gene Expression

Having a hard time deciding what controls to use to quantify gene expression—even with detailed information on biological systems? Now, with TaqMan™ endogenous controls, you can avoid all the trial-and-error of selecting controls for the most common human, mouse, rat, and eukaryotic genes.

Simple to Use

All components of the TaqMan™ endogenous controls are QC tested, formulated into a single 20X mix, and functionally tested. The controls are not only simple to use, but they are also fully compatible with universal conditions for two-step RT-PCR. Just add TaqMan™ Universal PCR Master Mix (with or without AmpErase™ UNG) and your cDNA sample to generate sensitive, reproducible, and truly quantitative gene expression data on Applied Biosystems instruments including the Applied Biosystems 7900HT, 7300, 7500 Real-Time PCR Systems, and the 7000 and 7700 Sequence Detection Systems.

Flexible Offering

We build each endogenous control using our proven 5' nuclease chemistry. For maximum flexibility, you can choose between two different reporter dyes and two quenchers:
  • A FAM™ dye-labeled TaqMan™ MGB probe (250nM, final concentration) and two unlabeled PCR primers (900nM each)
  • A VIC™ dye-labeled TaqMan™ MGB probe (250 nM, final concentration) and two unlabeled PCR primers (150 nM each, primer limited)
  • A VIC™ dye-labeled TAMRA™ probe (250nM, final concentration) and two unlabeled PCR primers (150 nM each, primer limited)

    Choosing the Right Endogenous Control

    Endogenous controls can normalize the expression levels of target genes by correcting differences in the amount of cDNA that is loaded into PCR reaction wells. For best results, verify that the endogenous control is consistently expressed in the sample set to be tested. Endogenous control expression must be uniform across all samples in the study. For multiplexing, ensure that the gene expression level of the endogenous control is greater than that of the target.

    Multiplex vs. Singleplex PCR

    All TaqMan™ endogenous controls that contain probes labeled with the VIC™ reporter dye are primer limited. This allows multiplexing of TaqMan™ endogenous controls with target gene expression assays, provided that the control gene is more abundantly expressed than the target gene. All TaqMan™ endogenous controls that contain probes labeled with the FAM™ reporter dye are not primer limited and are not intended for multiplexing.

  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage 4 tubes containing 20X mix of pre-formulated assay (1 probe and 2 primers), Control primer-probe set(s)
    –15 to –25°C
    Description Human RPLPO (Large Ribosomal Protein) Endogenous Control, 2500rxns, Tube(s) Format, VIC™ Dye Type, RPLPO Target Gene, Human Species, Format, –15 to –25°C Recommended Storage, For Real-Time PCR (qPCR), Real-Time PCR-Based Gene Expression Profiling
    Quantity 2500 rxn(s)
    For Use With (Application) Gene Expression

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.