missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ROR2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems AF2064-SP
This item is not returnable.
View return policy
Description
ROR2 Polyclonal specifically detects ROR2 in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| ROR2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BDB, BDB1, EC 2.7.10.1, MGC163394, neurotrophic tyrosine kinase receptor-related 2, Neurotrophic tyrosine kinase, receptor-related 2, NTRKR2, NTRKR2tyrosine-protein kinase transmembrane receptor ROR2, receptor tyrosine kinase-like orphan receptor 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4920 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| AAA60276 | |
| ROR2 | |
| Mouse myeloma cell line NS0-derived recombinant human ROR2 Val28-Gly403 Accession # AAA60276 | |
| 25 μg | |
| Primary | |
| Detects human ROR2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human ROR1 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction