missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ROR1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 13 publications
Brand: R&D Systems AF2000
This item is not returnable.
View return policy
Description
ROR1 Polyclonal specifically detects ROR1 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, CyTOF-ready.
Specifications
| ROR1 | |
| Polyclonal | |
| Unconjugated | |
| Q01973 | |
| ROR1 | |
| Mouse myeloma cell line NS0-derived recombinant human ROR1 Ala24-Glu403 Accession # Q01973 | |
| 100 μg | |
| Primary | |
| Detects human ROR1 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 25 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| EC 2.7.10.1, MGC99659, neurotrophic tyrosine kinase receptor-related 1, Neurotrophic tyrosine kinase, receptor-related 1, NTRKR1, NTRKR1dJ537F10.1, receptor tyrosine kinase-like orphan receptor 1, tyrosine-protein kinase transmembrane receptor ROR1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4919 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction