missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ROR alpha/NR1F1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8924
This item is not returnable.
View return policy
Description
ROR alpha/NR1F1 Monoclonal specifically detects ROR alpha/NR1F1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| ROR alpha/NR1F1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp686M2414, MGC119326, NR1F1, NR1F1MGC119329, nuclear receptor ROR-alpha, Nuclear receptor RZR-alpha, Nuclear receptor subfamily 1 group F member 1, RAR-related orphan receptor A, RAR-related orphan receptor alpha, retinoic acid receptor-related orphan receptor alpha, retinoid-related orphan receptor alpha, Retinoid-related orphan receptor-alpha, ROR1, ROR2, ROR3, RORA, RZRA, RZR-ALPHA, RZRAROR-alpha, transcription factor RZR-alpha | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6095 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 784652 | |
| Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| P35398 | |
| RORA | |
| E. coli-derived recombinant human ROR alpha/NR1F1 Arg47-His163 Accession # P35398 | |
| 100 μg | |
| Primary | |
| Detects human ROR alpha/NR1F1 in direct ELISAs. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction