missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ROR alpha/NR1F1 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC8924G-100UG
This item is not returnable.
View return policy
Description
ROR alpha/NR1F1 Monoclonal specifically detects ROR alpha/NR1F1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| ROR alpha/NR1F1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| DKFZp686M2414, MGC119326, NR1F1, NR1F1MGC119329, nuclear receptor ROR-alpha, Nuclear receptor RZR-alpha, Nuclear receptor subfamily 1 group F member 1, RAR-related orphan receptor A, RAR-related orphan receptor alpha, retinoic acid receptor-related orphan receptor alpha, retinoid-related orphan receptor alpha, Retinoid-related orphan receptor-alpha, ROR1, ROR2, ROR3, RORA, RZRA, RZR-ALPHA, RZRAROR-alpha, transcription factor RZR-alpha | |
| E. coli-derived recombinant human ROR alpha/NR1F1 Arg47-His163 Accession # P35398 | |
| RUO | |
| 6095 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 784652 | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P35398 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human ROR alpha/NR1F1 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction