missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ROBO4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2366-SP
This item is not returnable.
View return policy
Description
ROBO4 Polyclonal specifically detects ROBO4 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| ROBO4 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ECSM4, FLJ20798, Magic roundabout, MGC133352, MGC133353, MRB, roundabout homolog 4, roundabout homolog 4, magic roundabout (Drosophila) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 54538 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q8WZ75 | |
| ROBO4 | |
| Mouse myeloma cell line NS0-derived recombinant human ROBO4 Gln28-Arg467 Accession # Q8WZ75 | |
| 25 μg | |
| Primary | |
| Detects human ROBO4 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant rat ROBO1, recombinant human (rh) ROBO2, and rhROBO3 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction