missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RNF92 (aa 138-209) Control Fragment Recombinant Protein

Product Code. 30213368
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30213368

Brand: Invitrogen™ RP106054

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to the nucleus and the cytoplasm. Its function has not been identified.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96F44
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 81559
Name Human RNF92 (aa 138-209) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias BIA1; E3 ubiquitin-protein ligase TRIM11; Protein BIA1; RING finger protein 92; RING-type E3 ubiquitin transferase TRIM11; RNF92; Trim11; tripartite motif containing 11; tripartite motif protein 11; tripartite motif-containing 11; tripartite motif-containing protein 11
Common Name RNF92
Gene Symbol TRIM11
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KLEKSLEHLRKQMQDALLFQAQADETCVLWQKMVESQRQNVLGEFERLRRLLAEEEQQLLQRLEEEELEVLP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Vedi altri risultati Mostra meno risultati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato