missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RNF36 (aa 14-157) Control Fragment Recombinant Protein

Product Code. 30203559
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30203559

Brand: Invitrogen™ RP102305

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56006 (PA5-56006. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the RING-B-box-coiled-coil (RBCC) family and encodes a protein with an N-terminal RING finger motif, a PRY domain and a C-terminal SPRY domain. The mouse ortholog of this gene is specifically expressed in germ cells at the round spermatid stages during spermatogenesis and, when overexpressed, induces apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q86WT6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 140691
Name Human RNF36 (aa 14-157) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4921519C19Rik; E3 ubiquitin-protein ligase TRIM69; hCG_39321; HSD34; HSD-34; RFP-like (B30.2) domain protein; RFP-like domain-containing protein trimless; RING finger B-box coiled-coil transcription factor; ring finger protein 36; RING-type E3 ubiquitin transferase TRIM69; RNF36; testis-specific RING finger protein; Trif; Trim69; Trimless; tripartite motif containing 69; tripartite motif-containing 69; tripartite motif-containing protein 69
Common Name RNF36
Gene Symbol TRIM69
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GDYVEMNDSITHLPSKVVIQDITMELHCPLCNDWFRDPLMLSCGHNFCEACIQDFWRLQAKETFCPECKMLCQYNNCTFNPVLDKLVEKIKKLPLLKGHPQCPEHGENLKLFSKPDGKLICFQCKDARLSVGQSKEFLQISDAV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.