missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RNF23 (aa 85-152) Control Fragment Recombinant Protein

Product Code. 30181687
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30181687

Brand: Invitrogen™ RP99607

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62321 (PA5-62321. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The function of this protein has not been identified. This gene lies within the major histocompatibility complex class I region on chromosome 6. Alternate splicing results in two transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9HCM9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 56658
Name Human RNF23 (aa 85-152) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1100001D15Rik; E130103K13Rik; E3 ubiquitin-protein ligase TRIM39; mKIAA4179; RBCC-B30.2; ring finger protein 23; RING-B box-coiled coil-B30.2; RING-B box-coiled-coil-B30.2; RING-type E3 ubiquitin transferase TRIM39; Rnf23; Testis-abundant finger protein; tfp; TRIM39; TRIM39B; tripartite motif containing 39; tripartite motif protein 39; tripartite motif-containing 39; tripartite motif-containing protein 39
Common Name RNF23
Gene Symbol TRIM39
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GSMVEIAKQLQAVKRKIRDESLCPQHHEALSLFCYEDQEAVCLICAISHTHRAHTVVPLDDATQEYKE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.