missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RNF216 (aa 288-367) Control Fragment Recombinant Protein

Product Code. 30202630
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30202630

Brand: Invitrogen™ RP92071

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82649 (PA5-82649. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Activation of NF-κB as a result of Toll-like receptor (TLR) and IL-1 receptor signaling is a major component of innate immune responses. Signals from these receptors are relayed by a number of adapter molecules such as TRIF, TIRAP, and MyD88. Several regulatory mechanisms exist to control TLR signal transduction, including the inhibition of TLR expression and signaling by molecules such as ST2 and SIGIRR. Another mechanism is by the ubi-quitinization of selected TLRs by TRIAD3A, an E3 ubiquitin-protein ligase. TRIAD3A is a RING finger protein that can bind to TLR4 and TLR9, and to a lesser extent TLR3 and TLR5, catalyzing the ubiquitization of these molecules. Overexpression of TRIAD3A promoted the nearly complete degradation of TLR4 and TLR9; this reduction was reflected in the decreased signal-specific activation by ligands specific for these TLRs. Conversely, depletion of TRIAD3A resulted in enhanced TLR activation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NWF9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 54476
Name Human RNF216 (aa 288-367) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2810055G22Rik; AI647468; AU019462; C86502; CAHH; E3 ubiquitin ligase; E3 ubiquitin-protein ligase RNF216; F830018F18Rik; RGD1565135; RING finger protein 216; RING-type E3 ubiquitin transferase RNF216; RNF216; Triad domain-containing protein 3; TRIAD3; triad3a; U7I1; UBCE7IP1; UbcM4-interacting protein 83; ubiquitin-conjugating enzyme 7-interacting protein 1; Uip83; Zin; zinc finger protein inhibiting NF-kappa-B
Common Name RNF216
Gene Symbol RNF216
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VKETEARFPDVANGFIEEIIHFKNYYDLNVLCNFLLENPDYPKREDRIIINPSSSLLASQDETKLPKIDFFDYSKLTPLD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.