missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RNF17 Partial ORF (NP_112567, 30 a.a. - 139 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16178696
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16178696 10 μg 10µg
16188696 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16178696 Supplier Abnova™ Supplier No. H00056163Q01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

This gene is similar to a mouse gene that encodes a testis-specific protein containing a RING finger domain. [provided by RefSeq]

Sequence: IQCTRCGRRVSRSSGHHCELQCGHAFCELCLLMTEECTTIICPDCEVATAVNTRQRYYPMAGYIKEDSIMEKLQPKTIKNCSQDFKKTADQLTTGLERSASTDKTLLNSS

Specifications

Accession Number NP_112567
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 56163
Molecular Weight (g/mol) 37.84kDa
Name RNF17 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen IQCTRCGRRVSRSSGHHCELQCGHAFCELCLLMTEECTTIICPDCEVATAVNTRQRYYPMAGYIKEDSIMEKLQPKTIKNCSQDFKKTADQLTTGLERSASTDKTLLNSS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ11045/Mmip-2/SPATA23/TDRD4
Common Name RNF17
Gene Symbol RNF17
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.