missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RNF135 (aa 167-266) Control Fragment Recombinant Protein

Product Code. 30194863
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30194863

Brand: Invitrogen™ RP93486

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (46%), Rat (46%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54457 (PA5-54457. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by RNF135 contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. This gene is located in a chromosomal region known to be frequently deleted in patients with neurofibromatosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8IUD6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 84282
Name Human RNF135 (aa 167-266) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 0610037N03Rik; 2410006N06Rik; E3 ubiquitin-protein ligase RNF135; L13; LRRGT00184; MGC13061; MMFD; REUL; RIG-I E3 ubiquitin ligase; ring finger protein 135; RING finger protein 135; E3 ubiquitin-protein ligase RNF135; RING-type E3 ubiquitin transferase RNF135; Riplet; RNF135; U 2-3-0
Common Name RNF135
Gene Symbol RNF135
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LSILGKAFSSGVDLSMASPKLVTSDTAAGKIRDILHDLEEIQEKLQESVTWKEAPEAQMQGELLEAPSSSSCPLPDQSHPALRRASRFAQWAIHPTFNLK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.