missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RNASE3 (aa 82-136) Control Fragment Recombinant Protein

Product Code. 30207565
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30207565

Brand: Invitrogen™ RP95687

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (38%), Rat (38%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63260 (PA5-63260. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ECP (eosinophil cationic protein), also known as RNS3 or RNASE3 (ribonuclease, RNase A family, 3), is a 160 amino acid potent cytotoxic secretory protein that belongs to the pancreatic ribonuclease family. ECP localizes to the granule matrix of the eosinophil and is involved in the immune response system. ECP is a cytotoxin, neurotoxin and helminthotoxin that is secreted by activated human eosinophils. ECP exhibits anti-parasitic, antibacterial and ribonucleolytic activities. It has been suggested that ECP induces the neurotoxic effect known as the Gordon phenomenon, a syndrome manifested by ataxia, muscular rigidity, paralysis and tremor that may lead to death. ECP is considered a marker of eosinophilic inflammation and high levels have been found in cases of active asthma and other allergic diseases.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P12724
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6037
Name Human RNASE3 (aa 82-136) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias cytotoxic ribonuclease; Ear11; EAR-11; Ear3; ECP; Eosinophil cationic protein; eosinophil cationic protein-like; eosinophil cationic-type ribonuclease 3; Eosinophil secondary granule ribonuclease 11; eosinophil-associated ribonuclease 3; eosinophil-associated, ribonuclease A family, member 3; LOC100361866; mR3; MR-3; murine ribonuclease 3; RAF1; Ribonuclease 3; ribonuclease A family member 3; ribonuclease, RNase A family 3; ribonuclease, RNase A family, 3; RNase 3; RNASE3; RNS3
Common Name RNASE3
Gene Symbol RNASE3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence CGNQSIRCPHNRTLNNCHRSRFRVPLLHCDLINPGAQNISNCTYADRPGRRFYVV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.