missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Rit2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6094-SP
This item is not returnable.
View return policy
Description
Rit2 Monoclonal specifically detects Rit2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Rit2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Q99578 | |
| RIT2 | |
| E. coli-derived recombinant human Rit2 aa 1-217 Accession # Q99578 | |
| 25 μg | |
| Primary | |
| Detects human Rit2 in Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 456313 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GTP-binding protein Rit2, GTP-binding protein Roc2, Ras-like protein expressed in neurons, Ras-like without CAAX 2, Ras-like without CAAX protein 2, RIBA, Ric (Drosophila)-like, expressed in neurons, Ric-like, expressed in neurons, RIN, ROC2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6014 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction