missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Rit1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4697
This item is not returnable.
View return policy
Description
Rit1 Monoclonal specifically detects Rit1 in Human samples. It is validated for Western Blot.
Specifications
| Rit1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q92963 | |
| RIT1 | |
| E. coli-derived recombinant human Rit1 Met1-Thr219 Accession # Q92963 | |
| 100 μg | |
| Primary | |
| Detects human Rit1 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 456201 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GTP-binding protein Rit1, GTP-binding protein Roc1, MGC125864, MGC125865, Ras-like protein expressed in many tissues, Ras-like without CAAX 1, Ras-like without CAAX protein 1, RIBB, RIBBRIT, Ric-like, expressed in many tissues, RIT, ROC1, ROC1Ric (Drosophila)-like, expressed in many tissues | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6016 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction