missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RIM2 Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Rim2 Synthetic Peptide, PEP-0642, from Invitrogen.
Rab3-interacting molecules (RIMs) are synaptic proteins necessary for neuronal transmission and plasticity. Rim1 and Rim2 proteins are expressed in overlapping but distinct patterns throughout the brain. While the ablation of either gene was not lethal in mice, the deletion of both resulted in postnatal mortality. This lethality is due to a defect in neurotransmitter release; synapses without RIM proteins can still release neurotransmitters but are unable to do so in response to normal Ca2+ triggers. Like Rim1, Rim2 is thought to be an effector protein for Rab3, binding to Rab3 on synaptic vesicles in a GTP-dependent manner.
Specifications
Specifications
| Gene ID (Entrez) | 9699 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q9UQ26 |
| Common Name | Human RIM2 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction