missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Ribophorin II (aa 103-225) Control Fragment Recombinant Protein

Product Code. 30213415
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30213415

Brand: Invitrogen™ RP89457

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52540 (PA5-52540. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

RNP2 a type I integral membrane protein found only in the rough endoplasmic reticulum. The encoded protein is part of an N-oligosaccharyl transferase complex that links high mannose oligosaccharides to asparagine residues found in the Asn-X-Ser/Thr consensus motif of nascent polypeptide chains. This protein is similar in sequence to the yeast oligosaccharyl transferase subunit SWP1.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P04844
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6185
Name Human Ribophorin II (aa 103-225) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1300012C06Rik; AV261018; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase 63 kDa subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 2; hypothetical protein LOC534231; oligosaccharyltransferase complex subunit (non-catalytic); RIBIIR; ribophorin 2; ribophorin II; Ribophorin-2; RPN2; Rpn-2; RPNII; RPN-II; SWP1
Common Name Ribophorin II
Gene Symbol RPN2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SISNETKDLLLAAVSEDSSVTQIYHAVAALSGFGLPLASQEALSALTARLSKEETVLATVQALQTASHLSQQADLRSIVEEIEDLVARLDELGGVYLQFEEGLETTALFVAATYKLMDHVGTE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.