missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RhoF (aa 35-73) Control Fragment Recombinant Protein

Product Code. 30211543
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30211543

Brand: Invitrogen™ RP108276

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84989 (PA5-84989. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Plasma membrane-associated small GTPase which cycles between an active GTP-bound and an inactive GDP-bound state. Causes the formation of thin, actin-rich surface projections called filopodia. Functions cooperatively with CDC42 and Rac to generate additional structures, increasing the diversity of actin-based morphology.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9HBH0
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 54509
Name Human RhoF (aa 35-73) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI845056; Arhf; AV026554; Ifld1; induced in fatty liver dystrophy 1; ras homolog family member F (in filopodia); ras homolog family member F, filopodia associated; ras homolog gene family, member f; ras homolog gene family, member f (in filopodia); rho family GTPase Rif; Rho in filopodia; RHOF; rho-related GTP-binding protein RhoF; RIF
Common Name RhoF
Gene Symbol RHOF
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LLMVYSQGSFPEHYAPSVFEKYTASVTVGSKEVTLNLYD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.