missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RHCE (aa 388-417) Control Fragment Recombinant Protein

Product Code. 30209384
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30209384

Brand: Invitrogen™ RP109627

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (70%), Rat (70%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-140007 (PA5-140007. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

May be part of an oligomeric complex which is likely to have a transport or channel function in the erythrocyte membrane.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P18577
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6006
Name Human RHCE (aa 388-417) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias (C)ces type 1 Rhesus blood group D antigen; blood group Rh(CE) polypeptide; blood group RhCcEe antigen; blood group RhCE polypeptide; CD240CE; RH; Rh blood group antigen Evans; Rh blood group C antigen; Rh blood group CcEe antigens; Rh blood group protein; Rh blood group, CcEe antigens; rh polypeptide 1; Rh polypeptide I; Rh30A; Rh4; RHC; RHCE; RhCE blood group antigens; RHCE blood group variant Crawford antigen Rh43; RHE; rhesus blood group antigen, RhC antigen; Rhesus blood group CE protein; Rhesus blood group E antigen; rhesus blood group little e antigen; Rhesus blood group Rhce antigen; rhesus C/E antigens; Rhesus system C and E polypeptides; RhIVb(J); RHIXB; RhPI; RhVI; RhVIII; silenced Rh blood group CcEe antigen; truncated RhCcEe antigen; truncated RHCE
Common Name RHCE
Gene Symbol RHCE
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LNLKIWKAPHVAKYFDDQVFWKFPHLAVGF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.