missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human RGM-A Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2459-SP
This item is not returnable.
View return policy
Description
RGM-A Polyclonal specifically detects RGM-A in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| RGM-A | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| RGM domain family member A, RGM domain family, member A, RGMA, RGMrepulsive guidance molecule A | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 56963 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 10-25 ug/mL | |
| Q96B86 | |
| RGMA | |
| Mouse myeloma cell line NS0-derived recombinant human RGM-A Cys48-Gly422 Accession # Q96B86 | |
| 25 μg | |
| Primary | |
| Detects human RGM-A in direct ELISAs and Western blots. In direct ELISAs, less than 40% cross-reactivity with recombinant chicken RGM and recombinant mouse RGM-A is observed, and less than 1% cross-reactivity with recombinant human (rh) RGM-B and rhRGM-C is observed.. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction