missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RETSAT (aa 28-169) Control Fragment Recombinant Protein

Product Code. 30196243
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30196243

Brand: Invitrogen™ RP104822

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65443 (PA5-65443. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Retinol saturase carrying out the saturation of the 13-14 double bond of all-trans-retinol to produce all-trans-13,14-dihydroretinol. Has activity toward all-trans-retinol as substrate. Does not use all-trans-retinoic acid nor 9-cis, 11-cis or 13-cis-retinol isomers as substrates. May play a role in the metabolism of vitamin A.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6NUM9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 54884
Name Human RETSAT (aa 28-169) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias All-trans-13,14-dihydroretinol saturase; all-trans-retinol 13,14-reductase; PPAR-alpha-regulated and starvation-induced gene protein; PPSIG; retinol saturase; retinol saturase (all-trans-retinol 13,14-reductase); RETSAT; UNQ439/PRO872
Common Name RETSAT
Gene Symbol RETSAT
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SSPNPFSEDVKRPPAPLVTDKEARKKVLKQAFSANQVPEKLDVVVIGSGFGGLAAAAILAKAGKRVLVLEQHTKAGGCCHTFGKNGLEFDTGIHYIGRMEEGSIGRFILDQITEGQLDWAPLSSPFDIMVLEGPNGRKEYPM
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.