missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Respiratory Syncytial Virus Phosphoprotein Antibody (3-5-18), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50244DL594
340.00 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Human Respiratory Syncytial Virus Phosphoprotein Monoclonal antibody specifically detects Human Respiratory Syncytial Virus Phosphoprotein in Virus samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Human Respiratory Syncytial Virus Phosphoprotein | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| The immunogen was gradient-purified RSN-2 virus (subgroup B) that was then treated with 0.1% SDS at 100C for 2 min. The procedure used to produce this antibody is described in Gimenez et al. (1984). | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunofluorescence | |
| 2018-03-05 00:00:00 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction