missing translation for 'onlineSavingsMsg'
Learn More

Human Respiratory Syncytial Virus Nucleoprotein VPN41 Antibody (4-18), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18167678 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18167678 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18167678 Supplier Novus Biologicals Supplier No. NBP250535AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Human Respiratory Syncytial Virus Nucleoprotein VPN41 Monoclonal antibody specifically detects Human Respiratory Syncytial Virus Nucleoprotein VPN41 in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human Respiratory Syncytial Virus Nucleoprotein VPN41
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 43573
Conjugate Alexa Fluor 750
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Host Species Mouse
Immunogen Gradient-purified RSN-2 virus (subgroup B) treated with 0.1% SDS at 100C for 2 mins.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity This antibody reacts with both human RSV subgroups A and B.
Target Species Virus
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.